Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Bench-Top Biohazard Bag Holder
SCI_ED

Catalog No. S04788 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Product Type:
Bag Holder Cover
Biohazard Bag Holder
Biohazard Bag Holder Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S04788 Bag Holder Cover
S50397A Biohazard Bag Holder
S04788B Biohazard Bag Holder Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S04788 Supplier Bel-Art™ Supplier No. F131930102/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Plastic Cover Only

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use

  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28 cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812

Order Info

For complete kit with bag holder, cover and biohazard bags, see Cat. No. S04788B.

Specifications

Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With (Application) Benchtop biohazard bag holder
For Use With (Equipment) Biohazard bag holder
Includes Snug fitting plastic cover
Material Polyethylene
Dimensions (L x W x H) 12.7 x 8.9 x 1.6 cm (5 x 3-1/2 x 5/8 in.)
Width (English) 5 in.
Product Type Bag Holder Cover
Width (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.