Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Biohazard Sharp Object Safety Pouch Stand
GSA_VA

Catalog No. 22197061 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (English):
1 safety pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-197061 1 safety pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-197061 Supplier Bel-Art™ Supplier No. F132360000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Offers protection against cuts by contaminated sharp objects during work in labs, clinics, and medical offices

  • Provides a stable support to keep the Biohazard Sharp Object Safety Pouch ( Mfr. No. 13234-0000) upright and open for easy access
  • Constructed from bright orange, epoxy-coated steel wire
  • Frame dimensions are 10 x 13 x 20 cm H (4 x 5.125 x 8 in.)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity (English) 1 safety pouch
Color White
Autoclavable Not Autoclavable
For Use With (Application) For providing stable support to biohazard sharp objects safety pouch
Material Epoxy-coated Steel Wire
Product Type Sharps Container
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.