Learn More
Bel-Art™ Bellows Type Glove Box Gloves
Catalog No. p-172649 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Size:
Large (10)
Medium (9)
Small (8)
Product Type:
Bellows and Glove
Replacement Glove
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Product Type
11-391-105B Medium (9) Bellows and Glove
11-391-50 Small (8) Replacement Glove
11-391-52 Large (10) Replacement Glove
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings.
Catalog No. 11-391-105B Supplier Bel-Art™ Supplier No. H500250309
Products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings.
Forms a leak-proof unit with the box. Made from natural latex with a removable hand, the sleeves fit 20 cm (8 in.) circular ports and 18 x 25 cm (7 x 10 in.) oval ports.
  • Forms a leak-proof unit with O-ring construction integral with the box
  • Bellows allows cooling air circulation
  • When the glove and sleeve are separated, the sleeve remains attached to the box
  • Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
  • Length of the extended sleeve and glove is approximately 76 cm (30 in.)
  • Use with H50025-0320 Clamping Rings to hold gloves in place
  • Note: Due to the unspecified nature of their use and the environment in which they are used, glove box gloves may only be returned due to shipping damage, and only if they have not been used
Order Info
Sleeve fits 8 in. O.D. (20 cm) circular ports and 7 x 10 in. (18 x 25 cm) oval ports. Extended length with glove, 30 in. (76 cm). One pair per pack.
Specifications
Product Type Bellows and Glove
Material Natural Rubber, Neoprene
For Use With (Equipment) Glove box with 8 in. O.D. circular ports and 7 x 10 in. oval port
Size Medium (9)
WARNING: Cancer - www.P65Warnings.ca.gov