Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Cleanware™ Microscope Optics Cleaning Kit
SCI_ED

Catalog No. S193141 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Scent:
Unscented
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent
S193141 Unscented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S193141 Supplier Bel-Art™ Supplier No. F170720000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains everything you need to keep optics and microscope parts sanitary, clean and clear

  • Ten, pre-saturated optical cleaning pads
  • Two individually wrapped packs of anti-static plastic surface wipes
  • Two, 15.2cm (6 in.) swabs pre-moistened with alcohol(2)
  • Complete instructions for use

Specifications

Product Type Microscope Optics Cleaning Kit
Color-coded No
Form Unit Dose Concentrate
Scent Unscented
Sterility Non-sterile
Includes 10 Presaturated lens cleaning pads, two individually wrapped packs of anti-static Plastic surface wipes, two 6 in. (15.2cm) swabs, pre-moistened with germicide, lens tissue sold separately
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.