Learn More
Bel-Art™ Cloning Cylinders
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
Manufacturer:  Bel-Art™ F378470300
 View more versions of this product
Catalog No. 14-512-66
Add to cart
Description
Description
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Specifications
Specifications
0.37 in.
Individual clone isolation and propagation
11mm
Sterile
Cloning Cylinder
9.5mm
0.443 in.
Large
Polystyrene