Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Cryo-Safe™ Cold Box

Catalog No. 1451263 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
12 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-512-63 12 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-512-63 Supplier Bel-Art™ Supplier No. F188470002
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maintains the low temperature required for safely storing and transporting enzymes and other critical samples

  • Insulating gel keeps samples cold on the bench
  • Holds twelve 1.5 mL microcentrifuge tubes
  • Chill unit in a -20°C (-4°F) freezer
  • Maintains -15°C (+5°F) temperature for 1.5 hours
  • Remains below 0°C (32°F) for four hours
  • Weighs less than 1 lb. (0.45 kg)

Specifications

Height (Metric) 7.1 cm
Height (English) 2.7 in.
Length (Metric) 11.7 cm
Length (English) 4.6 in.
Width (Metric) 11.7 cm
Width (English) 4.6 in.
Dimensions (L x W x H) 4.6 x 4.6 x 2.7 in. (11.7 x 11.7 x 7.1 cm)
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Material Polystyrene
Product Type Insulated Container
Color White
Capacity (Metric) 12 tubes
Holds 12 (1.5mL microcentrifuge tube at -15°C temperature for 1.5 Hours and maintains 0°C for 4 Hours)
Autoclavable Not Autoclavable
Temperature Control No
Temperature Range -20°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.