Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Laboratory Tubing Fittings Assortment Kit
GSA_VASCI_ED

Catalog No. S90143 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S90143 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S90143 Supplier Bel-Art™ Supplier No. F196260000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fittings are for the most common lab connections and compatible with tubing measuring 3.2 to 12.7 mm.

  • Molded of autoclavable FDA grade polypropylene except quick disconnect fittings, which are polyethylene and not autoclavable or FDA grade
  • Kit contains: 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 “T” shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps
  • Plastic organizer box has room for adding other fittings and parts

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With 4.7 to 12.7mm
Includes 5 straight connectors, 4 stepped fittings, 8 Y fittings, 4 T connectors, 5 quick-disconnect fittings, 2 flow control tubing clamps
Color Natural
Product Type Fittings Assortment Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.