Learn More
Bel-Art™ SP Scienceware™ Fluorescent Ruler
GSA_VA
Catalog No. 0444016 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
04-440-16 Plastic
1 options
Catalog No. 04-440-16 Supplier Bel-Art™ Supplier No. F133550001
  • $66.20
    $55.65 / Each of 1
    Save $10.55
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Useful for all fluorescent viewing and fluorescent photographic work
  • The scale will glow visibly when illuminated with ultraviolet light
  • This Metric/English scale is printed on plastic with a pink phosphorescent ink
  • The scale is 0 to 15cm with millimeter increments on one edge and 0 to 6 in. divided into 0.0625 in. units on the other edge
  • Serves as a measure of nucleic acid mobility in gels
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Ruler, Fluorescent
Material Plastic