Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Glascribe™ Pen
GSA_VA

Catalog No. 22186171 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-186171 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-186171 Supplier Bel-Art™ Supplier No. H441500000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Permanently marks laboratory glassware, ceramics and plastics

  • Tungsten carbide tip (Mohs Hardness = 9) easily inscribes identifying marks on labware
  • Retractible tip operates with pen style push button
  • Clips to a pocket, ready for use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Scribe Pen
Tip Style Retractable
Color Black
Tip Material Tungsten Carbide
Includes Pocket clip
For Use With (Application) Permanently marks laboratory glassware, ceramics and plastics
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.