Learn More
Bel-Art™ Glove Box Economy Sleeved Gloves
Catalog No. 03421101 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
03-421-101 Black
1 options
This item is not returnable. View return policy
Catalog No. 03-421-101 Supplier Bel-Art™ Supplier No. H500290000
  • $817.00
    $567.65 / Pair of 1
    Save $249.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Compatible with Scienceware Portable Glove Box or any glove box with 6 in. glove ports
  • Durable neoprene fabrication
  • Safeguards against gas, liquid, or solid substance penetrants
  • Use with Clamping Ring, Part No. 03-421-102
  • Large, size 10
  • 1 pair/Pk.
Specifications
Color Black
Hand Specific
Latex Free No
Material Bonded Neoprene
Product Type Glove Box Gloves
Size 10
Thickness (English) 18 mil
Thickness (Metric) 0.4572 mm
WARNING: Cancer - www.P65Warnings.ca.gov