Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ HDPE Jerry Can with Spigot

Catalog No. 11865134 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-865-134 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-865-134 Supplier Bel-Art™ Supplier No. F118590010
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Jerry can features a smooth turning spigot that opens and closes with half a turn for easy dispensing from a shelf or rack.

  • Feature smooth turning spigot that opens and closes with half a turn for easy dispensing from a shelf or rack
  • An extra hand grip on bottom gives positive control when pouring
  • Graduated in U.S. gallons and liters, they have extra headspace for expansion of contents
  • 20 cm (8 in.) long spout with a built-in air vent for smooth pouring
  • Spout retracts into a collar sealed by a leakproof screw cap when not in use
  • 5L can with 2.5 cm (1 in.) I.D. pouring spout and 10 to 20L with 1.9 cm (0.75 in.) I.D. spouts

Specifications

Autoclavable Not Autoclavable
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material HDPE
Closure Type Screw Cap
Color White
For Use With (Application) Liquid Storage
Graduations Liters, U.S. gal.
Includes Spigot
Material HDPE
Mouth Style Wide
Product Type Jerry Can
Shape Rectangular
Sterility Non-sterile
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.