Learn More
Bel-Art™ SP Scienceware™ Lab-Aire II™ Double-Sided Electric Benchtop Glassware Dryers
SCI_ED
Catalog No. S93971 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Amperage:
8 A
Catalog No. Amperage
S93971 8 A
1 options
Catalog No. S93971 Supplier Bel-Art™ Supplier No. F188190023/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Offers compact footprint and accelerates drying of labware with warm, circulating air.
Double-sided models double the capacity with only a slightly larger footprint and can be turned perpendicular to the benchtop for easy access to both sides.
  • 400w of drying power gently circulates heated air into and around items to decreases drying time
  • Heat selector knob with three temperature settings: high, medium and low
  • Power button illuminates when in use
  • 8 Amp at 120V, circuit breaker and independent thermo-fuse protects unit from overheating
  • Efficiency is enhanced with replaceable polyurethane foam filter
  • Redirect air flow by inserting plugs instead of pegs into unused peg holes
  • Sturdy bases with excellent balance; can be placed on any flat surface
  • Removable polypropylene baskets for storage of small labware and extra pegs
  • 120V
  • Two or three tier option
  • Baskets hold small glassware including test tubes, pipets
Specifications
Material ABS
Product Type Double Sided Electric Dryer
Tiers 3 Tier
Includes 4 baskets, 76 pegs (24 small, 28 medium, 24 large) and 28 peg plugs
Amperage 8 A
Voltage 120 V