Learn More
Description
- Easily attached to all Bel-Art Lab-Aire™ II Dryers
- Helps keep gloves open for external and internal drying
- Extends glove life by maintaining shape, in addition to preventing bending and cracking
- Reduced drying time when attached to Electric Lab-Aire™ Glassware Drying racks, which circulate heated air in and around the gloves
- Epoxy-coated wire construction is resistant to chemicals and corrosion
- Holds one pair of gloves
Specifications
Specifications
| Color | Blue |
| Disposable | Reusable |
| Material | Epoxy-coated Wire |
| Product Type | Glove Clip |
| For Use With (Application) | Hanging gloves to dry, and maintaining glove shape |
| For Use With (Equipment) | Lab-Aire™ II Drying Racks |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.