Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Safety Labeled Wide-Mouth Wash Bottle
GREENER_CHOICESCI_ED

Catalog No. S90133 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S90133 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90133 Supplier Bel-Art™ Supplier No. F116462232/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information.

  • Safety labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information.
  • Easy-to-read printed bottles.
  • Wide mouth 53 mm (2 in.) bottle for easy refilling.
  • Manufactured from translucent, LDPE solvent-resistant.
  • Polypropylene closures.
  • Compliant with OSHA 29 CFR 1910.1200.

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Color Translucent, Red
For Use With (Application) Acetone
Material Polyethylene
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Chemical Label Acetone
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 32 oz.
Shape Cylindrical
Disposable Reusable
Product Type Safety-labeled Wash Bottle
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.