Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Labeled Wash Bottles
GREENER_CHOICESCI_ED

Catalog No. S66410 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
500 mL
For Use With (Application):
Isopropanol
Variety pack that includes: Water, Acetone, Methanol, Isopropanol, Toluene and Ethanol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) For Use With (Application)
S66410 500 mL Isopropanol
S317328 500 mL Variety pack that includes: Water, Acetone, Methanol, Isopropanol, Toluene and Ethanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.
Catalog No. S66410
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.

Includes

Safety labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemmical Abstract Service (CAS), protective equipment symbols and target organ information.

Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.

  • Easy-to-read printed bottles
  • Wide mouth 53 mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant
  • Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
  • Toluene is red LDPE bottle
  • Polypropylene closure with leakproof design
  • Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol

Compliance

OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Color Natural
For Use With (Application) Isopropanol
Material LDPE, Polyethylene
Sterility Non-sterile
Closure Color Yellow
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Chemical Label Isopropanol
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Cylindrical
Disposable Reusable
Product Type Safety-labeled Wash Bottle
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.