Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Lens Tissues

Catalog No. 1199750 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Bristle Material:
Cellulose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bristle Material
11-997-50 Cellulose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-997-50 Supplier Bel-Art™ Supplier No. F248410000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Silicone-free

  • For glass and plastic lenses
  • Pop-up dispenser pack
  • Sheet: 4.33 x 5 in. (108 x 127mm)

Specifications

Product Type Lens Cleaning Tissue
Bristle Material Cellulose
Bristle Type Paper
Color-coded No
Form Clear Liquid
pH 7
Scent Unscented
Sterility Non-sterile
For Use With (Application) Used to quickly and easily clean Glass and Plastic lenses
Length (Metric) 10.8 cm
Material Lint-free Tissue
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.