Learn More
Bel-Art™ Personal Desktop Standard Incubator
Catalog No. 03410542 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
No. of Places:
6
Catalog No. No. of Places
03-410-542 6
1 options
Catalog No. 03-410-542 Supplier Bel-Art™ Supplier No. 420900000
Incubate directly on your desktop
  • Small footprint
  • Ideal for Cell Culture
  • Keep samples seperated from others in the lab
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 1 cm3
Chamber Material Aluminum
Convection Type Forced
Material ABS Plastic
No. of Places 6
No. of Shelves 1
Plug Type CH
Relative Humidity 80%
Type Benchtop Incubator
Voltage 0/1 V