Learn More
Bel-Art™ Piezoelectric Gas Lighter
SCI_ED
Catalog No. S41878 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
For Use With (Equipment):
Flammable Items
Catalog No. For Use With (Equipment)
S41878 Flammable Items
1 options
This item is not returnable. View return policy
Catalog No. S41878 Supplier Bel-Art™ Supplier No. H150560000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Plastic handle and trigger with polished aluminum barrel.
  • Just squeeze the trigger for a hot ignition spark
  • The long barrel protects you by letting you light the fuel from a safe distance
  • Lighter operates without flint, fuel or batteries
  • The charge is induced by solid-state piezoelectric principle
Specifications
For Use With (Equipment) Flammable Items
Length (English) 10.438 in.
Length (Metric) 26.5 cm
Length (Metric) Blade 6 in.
Material Plastic
For Use With (Application) Plastic handle and trigger with polished aluminum barrel
Includes 5 Flints
Product Type Piezoelectric Gas Lighter