Learn More
Bel-Art™ Plate Reader
Catalog No. 14809175 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
14-809-175 None
1 options
Catalog No. 14-809-175 Supplier Bel-Art™ Supplier No. F378610000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Tool allows safe examination of the bottom of plates without risking spills on the face and eyes
  • For examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size
  • Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes
Hemagglutination, Hemagglutination Inhibition, Hemolysis and Aglutination testing
Specifications
Product Type Microtiter Plate Reader
Display Type None
Magnification 3X
Material Polypropylene, Clear Glass Acrylic Deck
For Use With (Application) Hemagglutination, hemagglutination inhibition, hemolysis and agglutination testing
For Use With (Equipment) Microtiter plates