Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Replacement Caps for Polypropylene Mason Jar Wide-Mouth Bottles

Catalog No. 22260150 Shop All SP Bel-Art & SP Wilmad Products
This item is not returnable. View return policy
Catalog No. 22-260150 Supplier Bel-Art™ Supplier No. F126400000

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heavy-duty, leakproof storage.

  • Closures feature handy grip lugs

Specifications

Color Translucent
Material Polypropylene
Sterility Non-sterile
Neck Finish 53-415
For Use With (Equipment) Polypropylene Mason Jar Wide-mouth Bottles
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Screw Cap
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.