Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Poxygrid™ Tube Slant Rack
GSA_VA

Catalog No. 1371721 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Positions:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
13-717-21 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-717-21 Supplier Bel-Art™ Supplier No. F187940028
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.

  • Holds eight 50mL conical tubes or other vessels up to 30mm in diameter
  • Can be attached to the underside of a shelf using built in mounting points (hardware not included); keeps samples off the bench but immediately available
  • Steel wire is epoxy coated for long life and easy cleaning; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Autoclavable at 121°C (250°F)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Not Autoclavable
Color Blue
Holds Tubes (30 mm), Tubes (5 mL)
Material Epoxy-coated Steel
Positions 8
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Conical tubes or other vessels up to 30mm dia.
Height (English) 0.75 in.
Diameter (Metric) Opening 30 mm
Product Type Tube Rack
Shelves 3 Tiers
Height (Metric) 120 cm
Array 2 x 4
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.