Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Poxygrid™ Wire Test Tube Racks, Holds Tubes 20 to 24 mm

Catalog No. 22260569 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds:
Tubes (20 to 24 mm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-260569 Tubes (20 to 24 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-260569 Supplier Bel-Art™ Supplier No. F187670001
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores 40 tubes
  • Autoclavable

Specifications

Autoclavable Autoclavable
Color Blue
Holds Tubes (20 to 24 mm)
Material Epoxy-coated Steel
Positions 40
Shape Rectangular
Stackable No
Sterility Non-sterile
Dimensions (L x W x H) 10.43 x 4.57 x 3.25 in. (26.49 x 11.6 x 8.3cm)
Length (Metric) 26.3 cm
Length (English) 10.35 in.
Width (English) 4.57 in.
Height (English) 3.25 in.
Product Type Tube Rack
Shelves None
Height (Metric) 8.3 cm
Array 4 x 10
Width (Metric) 11.6 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.