Learn More
Bel-Art™ PTFE Kit
Catalog No. 1116377A Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
For Use With (Equipment):
Flowmeter Frame
Catalog No. For Use With (Equipment)
11-163-77A Flowmeter Frame
1 options
Catalog No. 11-163-77A Supplier Bel-Art™ Supplier No. 404080000
Includes
PTFE frame with 3/8 in. O.D. (9.5mm) glass tube extensions; leveling base; four flowmeters (sizes 1, 2, 3, and 4); two support brackets; two mounting rods; two rod clamps; nine floats; cleaning kit; and carrying case.
Fluids contact PTFE fittings, float and glass Flow Tube
  • Range: 0.03 to 742mL/min. for water; 2.5 to 23,411mL/min. for air
Specifications
Voltage 0/1 V
For Use With (Equipment) Flowmeter Frame
Type Flow Indicator