Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Rack Slanter
GSA_VASCI_ED

Catalog No. s98144 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S98144 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S98144 Supplier Bel-Art™ Supplier No. F188900100/EMD
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tilts your test tube rack to 25°, 35° or 45°.

  • Adjusts the angle of racks to reduce hand strain when working with pipetters.
  • Fits racks up to 11.56 x 9.56 x 3.25 in.H (293 x 242 x 83 mmH)
  • Rigid polypropylene construction
  • Great for making perfectly angled agar slant tubes
  • Steam autoclavable at 121°C (250°F)
  • Also for use with the Mini-Magnifier to aid in the viewing of Petri dishes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Autoclavable
Color Translucent
Holds Test Tubes
Material Polypropylene
Positions 3
Shape Rectangular
Stackable No
Sterility Non-sterile
Dimensions (L x W x H) 11.56 x 9.56 x 3.25 in. (29.3 x 24.2 x 8.3cm)
Length (Metric) 29.3 cm
Length (English) 11.56 in.
Width (English) 9.56 in.
Height (English) 3.25 in.
Product Type Test Tube Slant Rack
Shelves None
Height (Metric) 8.3 cm
Array 1 x 1
Width (Metric) 24.2 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.