Description
- For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
- Pipettes fit smoothly into these flexible tapered chucks
- Replacement chucks to fit various sizes/colors of Pipette Pumps in case of loss or damage
- Silicone rubber is resistant to acids and alkalis
- Disassemble parts for cleaning; chuck should not be autoclaved
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With | Pipette Pump Pipettors |
| Material | Silicone Rubber |
| Type | Chuck |
| For Use With (Equipment) | 25mL pump pipettor |