Learn More
Bel-Art™ SP Scienceware™ Riteflow™ Flowmeter Leveling Base Kit
Catalog No. 1116380 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
For Use With (Application):
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
Catalog No. For Use With (Application)
11-163-80 Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
1 options
Catalog No. 11-163-80 Supplier Bel-Art™ Supplier No. H404041000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
  • 15.2cm (6 in.) Plexiglas™ triangular base
  • Three adjustable feet
  • Vial level
  • For use with Riteflow mounted flowmeters
Specifications
For Use With (Equipment) Scienceware Flowmeters Catalog Nos.: 11-163-75 and -77 Series
Type Leveling Base Kit
For Use With (Application) Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
Size 15.2 cm (6 in.)