Learn More
Bel-Art™ Safety Vinyl Goggles
Catalog No. 1451214 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
14-512-14 Clear
1 options
This item is not returnable. View return policy
Catalog No. 14-512-14 Supplier Bel-Art™ Supplier No. H248730000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits comfortably over prescription glasses.
  • Made of soft, clear, flexible vinyl
  • Conforms to face and nose
  • With 4 vents to reduce fogging
  • Clear polycarbonate lenses
  • Adjustable 13mm W (0.5 in.) elastic headband
Specifications
Lens Coating Uncoated
Lens Tint Clear
Product Type Safety Goggles
For Use With (Equipment) Prescription glasses
Includes Elastic headband
Material Vinyl
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
UV Protection None
Frame Type Contoured