Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ Secador™ Auto Desiccator Cabinet, Vertical Model
GSA_VA

Catalog No. p-4901863 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Color:
Amber
Blue with Clear Door
Clear
Clear with Blue Ends
Cabinet Style:
Vertical
Vertical (Tinted)
Voltage:
120 V
230 V
Volume (English):
1.16 cu. ft.
1.55 cu. ft.
1.87 cu. ft.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Cabinet Style Voltage Volume (English)
08-647-52 Clear Vertical 120 V 1.87 cu. ft.
08-647-655 Clear Vertical 120 V 1.55 cu. ft.
08-647-656 Clear Vertical 120 V 1.16 cu. ft.
08-647-668 Clear Vertical 230 V 1.55 cu. ft.
08-647-855 - Vertical (Tinted) 120 V 1.55 cu. ft.
08-647-858 Amber Vertical (Tinted) 120 V 1.87 cu. ft.
08-647-53 Clear with Blue Ends Vertical 120 V 1.87 cu. ft.
08-647-679 Blue with Clear Door Vertical 120 V 1.87 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 08-647-52 Supplier Bel Art™ Supplier No. F420741115
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A UV light blocking, dry environment with automatic electronic dehumidification.

A UV light blocking, dry environment with automatic electronic dehumidification.

  • Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
  • 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Order Info

Replacement shelves are available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Not Autoclavable
Color Clear
Cabinet Style Vertical
Material Copolyester
Product Type Desiccator
Door Style Side-swinging
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 4.0
No. of Shelves 3
Resistance Chemicals, stains and fine cracks
Shelf Type Vented Adjustable
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5°C to 40°C
Volume (English) 1.87 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.5 cm
Height (English) 20.4 in.
Height (English) Opening 16.4 in.
Height (Metric) 51.8 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Voltage 120 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.