Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Spill Containment Tray with Grid
GSA_VA

Catalog No. 0340950 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (English):
236.69 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
03-409-50 236.69 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-409-50 Supplier Bel-Art™ Supplier No. F246760003
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Large, deep, polypropylene tray protects users by capturing accidental hazardous spills

  • The epoxy coated wire support grid holds numerous vessels or racks of test tubes
  • Outstanding chemical resistance
  • Steam autoclavable at 121°C (250°F)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity (English) 236.69 oz.
Capacity (Metric) 7 L
Material Polypropylene
Type Spill Containment Tray
For Use With (Application) Bench top use
For Use With (Equipment) Multiple flasks, beakers, or a rack of test tubes
Includes Grid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.