Learn More
Description
- Polygon magnetic stirring bar spins within polystyrene capsule
- Prevents damage during pH adjustments and to gels during staining/destaining procedures
- Large, flat capsule keeps stirring bar consistently centered on bottom of vertical electrophoresis units to efficiently mix the running buffer
- Prevents stirring bar from being impeded by chemicals which, in water, transform into sticky gel and enables efficient dissolving
- After use, capsule can be easily disassembled for cleaning
- Autoclavable
Specifications
Specifications
| Type | Magnetic Stir Bar |
| Coating | PTFE, FDA-grade, USP Class VI |
| Diameter (Metric) | 5.3 cm |
| Length (Metric) | 5.3 cm |
| Quantity | 1 |
| Autoclavable | Autoclavable |
| Material | Polystyrene |
| Shape | Polygon |
| Includes | 30 x 8mm polygon magnetic stirring bar |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.