Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Spinbar™ Magnetic Stirring Bar Restrainer
SCI_ED

Catalog No. S96720 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Length (Metric):
8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S96720 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S96720 Supplier Bel-Art™ Supplier No. F377861000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fits ergonomically in one hand and when placed alongside the outer wall of a mixing vessel, it draws in and secures the stirring bar in situ.

  • Eliminate hazard caused with stir bar splashes when decanting mixing vessels
  • Comfortable, one-handed use
  • Fabricated with simple-to-clean durable polystyrene
  • Houses strong magnet that restrains magnetic stirring bars up to 80 mm (3 in.) long

Specifications

Dimensions (L x W x H) 8.71 x 2.87 x 4.24 cm
Diameter (Metric) 8.71 cm
Material Polystyrene, Magnetic Alloy
Quantity 1
Shape Rectangle
Type Stir Bar Retriever
Length (Metric) 8 cm
Length (English) 3 in.
For Use With (Application) Eliminate hazard caused with stir bar splashes when decanting mixing vessels, capture bars in containers and restrains bars up to 3 in. long
For Use With (Equipment) Stirring bars
Description Fabricated with simple-to-clean durable polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.