Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Standards Bottle Rack

Catalog No. 1371720 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds:
Vials (25 mm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
13-717-20 Vials (25 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-717-20 Supplier Bel-Art™ Supplier No. F185132300

missing translation for 'validMainframeMsg'

Organizes valuable USP™ reference standards bottles for convenient storage and access

  • Suitable for use in Secador™ 1.0 to 4.0 desiccators for low humidity storage
  • Clear acrylic construction for easy viewing of labels
  • Stores thirty 23mm standards bottles in a compact footprint

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Vials (25 mm)
Material Acrylic
Positions 30
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 13 x 3.37 x 0.87 in. (33 x 11 x 2.2cm)
Length (Metric) 33 cm
Length (English) 13 in.
Width (English) 3.37 in.
Height (English) 0.87 in.
Product Type Bottle Rack
Diameter (Metric) Inner 25 mm
Shelves 2 Tiers
Height (Metric) 2.2 cm
Array 10 x 3
Width (Metric) 11 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.