Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Switch-Grid™ Test Tube Rack, Rack Only
SCI_ED

Catalog No. S43179 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds:
Test Tube Rack Grids
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
S43179 Test Tube Rack Grids
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43179 Supplier Bel-Art™ Supplier No. F187450045/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bel-Art™ Switch-Grid™ rack transforms your lab with interchangeable grids, holding 2 tube sizes at once (10 to 30 mm). This is the rack only, intended for use with spare grids.

The innovative Switch-Grid rack utilizes interchangeable grids for ultimate flexibility. This split-frame rack holds up to two different tube sizes at the same time and allows users to quickly switch grids to hold other sizes as needed. This rack is a smart, adaptable solution for optimizing test tube organization and streamlining your lab's workflow.

  • 10 mm to 30 mm diameter test tubes fit in 5 color-coded grid inserts
  • Constructed from chemical-resistant polyoxymethylene plastic, both the frame and grids are autoclavable at 121°C (250°F)
  • Can be easily cleaned with common lab detergents, ethanol, or isopropyl alcohol
  • Three-tier design securely holds tubes in place while non-slip rubber feet prevent sliding on the lab bench
  • Rack available without grids for use with spare grids

Order Info

Grid sets in alternate sizes can be purchased separately.

Specifications

Autoclavable Autoclavable
Color White
Holds Test Tube Rack Grids
Material Plastic
Positions 2 x 1
Shape Rectangular, Rounded Corners
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) Switch-Grid Test Tube Rack
No. of Racks 1
Size L x W x H: 31.8 x 11.1 x 6.7 cm
Length (Metric) 320 mm
Product Type Switch Grid Test Tube Rack
Shelves 3 Tiers
Height (Metric) 70 mm
Array 2 x 1
Width (Metric) 112 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.