Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Three-In-One Reservoir

Catalog No. 0340952 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-409-52 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-409-52 Supplier Bel-Art™ Supplier No. F378820000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for use when filling multi-tube blocks or microwell plates, this two-sided polypropylene reservoir is three reservoirs in one

  • A graduated single reagent reservoir with graduations at 10, 25, 40 and 50mL
  • Reverse side has a reservoir with 8 separate 5mL (max. capacity) reagent channels and a reservoir with 12 separate 5mL (max. capacity) reagent channels
  • Each compartment of the multi-channel reservoirs is numbered for reagent identification
  • Suitable for use with all brands of multichannel pipettors
  • Autoclavable and reusable

Specifications

Color White
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Sterile
Type Pipetting Reservoir
For Use With (Application) Pipetting
Packaging Bag
Quantity 21
Autoclavable Autoclavable
Channels 8 and 12
Disposable Reusable
Bottom Shape V-bottom
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.