Learn More
Bel-Art™ SP Scienceware™ Vernier Calipers with Double Scales to 150mm
Catalog No. 12120 Shop All SP Bel-Art & SP Wilmad Products
Catalog No. 12-120 Supplier Bel-Art™ Supplier No. H134150000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Vernier calipers can be used to measure internal and external dimensions with utmost precision.
  • Precise measurements can be made with the fine adjustment thumb roller, and reading scales create no parallax
  • Parallel jaws are 1.55 in. (40mm) deep
  • Inside jaws and round plastic rod depth gauge provide added value
  • Upper scale: 150 mm with 0.1mm subdivisions; lower scale: 6 in. with 0.005 in. subdivisions
  • Molded in rigid plastic; will not scratch like metal
  • Non-conducting, corrosion resistant, and lightweight
Specifications
Product Type Caliper, Vernier
Material Polymeric Plastic