Learn More
Bel-Art™ Wet Surface Pen
Catalog No. 03422600 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
03-422-600 Each
1 options
Catalog No. 03-422-600 Supplier Bel-Art™ Supplier No. F133820000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Versatile pen writes clearly on wet, greasy, and other hard to mark surfaces
  • For use under water, upside down and in harsh temperatures
  • Writes on dry paper as a standard ballpoint pen
  • Excellent for marking slots on Southern, Northern and Western blots or for marking alignment dots on blotting filters placed over agar plates containing bacterial colonies or phage plaques
  • Mark lanes on wet blots prior to cutting the filter into strips to be incubated separately with different antisera or probes
  • Can also be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at -20°C (-4°F)
  • Extremely useful for marking filters during processing underwater-based buffers
  • Handy and reliable for writing on vertical surfaces
Order Info
Compatible with the Bel-Art Scienceware Economy Write-on™ Label Tape Dispenser Wall Mount Adapter (Mfr. No. 13473-0001), sold separately.
Specifications
Tip Style Rollerball
Product Type Wet Surface Pen
Color Black
For Use With (Application) Uses include changing information on wet labels, whether from water bath liquid nitrogen or a freezer. Mark slots on wet southern, northern and western blots
Quantity Each
WARNING: Cancer - www.P65Warnings.ca.gov