Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Y Connectors

Catalog No. 22235556 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
For Use With:
6mm
9.5mm
50mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
22-235556 50mm
22-235549 9.5mm
22-235531 6mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-235556 Supplier Bel-Art™ Supplier No. F196180000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)

  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable

Specifications

Autoclavable Autoclavable
Diameter (English) Inner 0.5 in.
Material Polypropylene
For Use With 50mm
Shape Y
Color Natural
Product Type Y Tubing Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.