Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Bulb Pipettor

Catalog No. 10987051 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Number of Channels:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
10-987-051 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-987-051 Supplier Bel-Art™ Supplier No. H378880000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bulb pipettor is an ideal aspirator for use with Westergren or Wintrobe sedimentation tubes. Allows users to keep the bulb with a reagant bottle.

  • Comes complete with an elastic cord for dedicating that pipettor to a specific reagent bottle
  • Tapered, silicone rubber seal provides an airtight fit on all size pipettes
  • Can be opened, cleaned and steam autoclaved at 121ºC (250ºF)

Specifications

Autoclavable Autoclavable
Color Red
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Rubber
Model Standard
Number of Channels 1
Sterility Non-sterile
Type Bulb Pipettor
For Use With (Equipment) Westergren and Wintrobe sedimentation tubes
Includes Elastic Cord
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.