Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Draft Checker Airflow Detection Powder

Catalog No. 10029252 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Quantity:
1.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-029-252 1.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-029-252 Supplier Bel-Art™ Supplier No. F199390000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A quick and easy way to test the safety of fume hood exhaust before use

Just a squeeze or two from the plastic bottle dispenses a cloud of ultra-fine, lightweight silica powder, which remains suspended long enough to visually check whether or not the fume hood exhaust is functioning properly.

  • Useful for detecting drafts, gas leaks and air currents in rooms, ducts and exhaust systems
  • Amorphous silica powder is inert, nontoxic and nonflammable
  • 1.5g bottle

Specifications

Description Airflow Detection Powder
Voltage 0/1 V
For Use With (Equipment) Flowmeter
Type Leak Detector
For Use With (Application) For testing the safety of fume hood exhaust before use.
Includes 1.5 g silica powder
Material Silica Powder
Quantity 1.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.