Learn More
MSC Bel-Art Wide-Mouth Bottle, Polyethylene, 16 oz, Translucent
Catalog No. 5000534798
Encompass
This wide-mouth bottle is constructed from durable polyethylene, making it suitable for a variety of laboratory applications. Its translucent design allows for convenient viewing of contents, and it features a generous volume capacity range of 16 to 31.9 oz.
  • Wide-mouth design for easy filling and dispensing.
  • Constructed from polyethylene for durability.
  • Translucent color provides clear visibility of contents.
  • Offers a capacity range of 16 to 31.9 oz.
Catalog No. 50-005-34798 Supplier MSC Supplier No. 65362634
This wide-mouth bottle is constructed from durable polyethylene, making it suitable for a variety of laboratory applications. Its translucent design allows for convenient viewing of contents, and it features a generous volume capacity range of 16 to 31.9 oz.
  • Wide-mouth design for easy filling and dispensing.
  • Constructed from polyethylene for durability.
  • Translucent color provides clear visibility of contents.
  • Offers a capacity range of 16 to 31.9 oz.