Learn More
Research Products International Corp Bel-Blotter 96 Well Replicating Tool, 8 x 12 Format
SDP
Catalog No. 50212586
Encompass
The Bel-Blotter 96 Well Replicating Tool quickly and conveniently duplicates 96 well plates. The blotter features unique 'open ended' tips that pick-up to 10ul of fluid per tip. Ideal for procedures such as recombinant DNA replication, colony hybridization, phage typing and more. Polycarbonate blotter fits all types of plates including flat, round and 'V' shaped bottom. Also suitable for replicating contents of micro-tubes when arranged in a 8 x 12 format.
Catalog No. 50-212-586 Supplier Research Products International Corp Supplier No. 248675
The Bel-Blotter 96 Well Replicating Tool quickly and conveniently duplicates 96 well plates. The blotter features unique 'open ended' tips that pick-up to 10ul of fluid per tip. Ideal for procedures such as recombinant DNA replication, colony hybridization, phage typing and more. Polycarbonate blotter fits all types of plates including flat, round and 'V' shaped bottom. Also suitable for replicating contents of micro-tubes when arranged in a 8 x 12 format.