Learn More
Cayman Chemical Irigenin, Purity: ≥98%, CAS: 548-76-5, 25 mg
SDP
Catalog No. 502950255
Encompass_Preferred
An isoflavonoid with diverse biological activities; inhibits CYP1A (IC50 = 1.2 µM); induces a 2-fold increase in NADPH quinone reductase activity in Hepa-1c1c7 cells at 7.8 µM; inhibits LPS-induced production of NO and PGE2, as well as increases in the levels of iNOS and COX-2, in RAW 264.7 cells; increases survival and reduces cardiac apoptosis, fibrosis, and levels of TNF-α, IL-6, IL-18, and IL-1β in a mouse model of doxorubicin-induced cardiotoxicity at 10 and 20 mg/kg
Catalog No. 50-295-0255 Supplier Cayman Chemical Supplier No. 3467625 MG
An isoflavonoid with diverse biological activities; inhibits CYP1A (IC50 = 1.2 µM); induces a 2-fold increase in NADPH quinone reductase activity in Hepa-1c1c7 cells at 7.8 µM; inhibits LPS-induced production of NO and PGE2, as well as increases in the levels of iNOS and COX-2, in RAW 264.7 cells; increases survival and reduces cardiac apoptosis, fibrosis, and levels of TNF-α, IL-6, IL-18, and IL-1β in a mouse model of doxorubicin-induced cardiotoxicity at 10 and 20 mg/kg