Learn More
Medchemexpress LLC HY-10225 200mg , Belinostat CAS:866323-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10225200MG
Encompass_Preferred
HY-10225 200mg Belinostat CAS: 866323-14-0 Belinostat (PXD101; PX105684) is a potent HDAC inhibitor with an IC50 of 27 nM in HeLa cell extracts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9294