Learn More
TARGETMOL CHEMICALS INC BELINOSTAT 50MG
SDP
Catalog No. 502579062
Encompass_Preferred
Also available in 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belinostat (PXD101) is an inhibitor of hydroxamic acid-type histone deacetylase (HDAC IC50 of 27 nM in HeLa cell extracts)and with antineoplastic activity. purity: 100%
Catalog No. 50-257-9062 Supplier TARGETMOL CHEMICALS INC Supplier No. T851750MG
Also available in 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belinostat (PXD101) is an inhibitor of hydroxamic acid-type histone deacetylase (HDAC IC50 of 27 nM in HeLa cell extracts)and with antineoplastic activity. purity: 100%