Learn More
Apexbio Technology LLC Belinostat (PXD101) 414864-00-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064357
Encompass_Preferred
Belinostat (PXD101) is a hydroxamic acid-based inhibitor targeting histone deacetylase (HDAC) enzymes Its mechanism involves the suppression of HDAC enzymatic activity leading to increased acetylation of histone proteins H3 and H4 within cells In biochemical assays using Hela cell extracts belinostat exhibits inhibitory activity against HDAC with an IC50 value of approximately 27 nM In tumor cell culture models belinostat induces cytotoxicity and represses cellular proliferation with reported IC50 values ranging roughly from 0 5 M to 10 M varying by cell type and origin making it suitable for research on epigenetic modulation in oncology specifically in studies of bladder prostate and related cancer cell lines
Catalog No. 50-000-64357 Supplier Apexbio Technology LLC Supplier No. A40965.1
Belinostat (PXD101) is a hydroxamic acid-based inhibitor targeting histone deacetylase (HDAC) enzymes Its mechanism involves the suppression of HDAC enzymatic activity leading to increased acetylation of histone proteins H3 and H4 within cells In biochemical assays using Hela cell extracts belinostat exhibits inhibitory activity against HDAC with an IC50 value of approximately 27 nM In tumor cell culture models belinostat induces cytotoxicity and represses cellular proliferation with reported IC50 values ranging roughly from 0 5 M to 10 M varying by cell type and origin making it suitable for research on epigenetic modulation in oncology specifically in studies of bladder prostate and related cancer cell lines