Learn More
Selleck Chemical LLC Belinostat (PXD101) 10mg 414864-00-9 NSC726630, PX-105684
SDP
Supplier:  Selleck Chemical LLC S108510MG
Encompass_Preferred
Belinostat (PXD101, NSC726630, PX-105684) is a novel HDAC inhibitor with IC50 of 27 nM in a cell-free assay, with activity demonstrated in cisplatin-resistant tumors. Belinostat (PXD101) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-835-1