Learn More
TARGETMOL CHEMICALS INC BELIZATINIB 10MG
SDP
Catalog No. 502579918
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belizatinib (TSR-011) is an effective oral and dual inhibitor of ALK (IC50 0.7 nM wild-type recombinant ALK) and TRKA TRKB and TRKC. purity: 99%
Catalog No. 50-257-9918 Supplier TARGETMOL CHEMICALS INC Supplier No. T425710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Belizatinib (TSR-011) is an effective oral and dual inhibitor of ALK (IC50 0.7 nM wild-type recombinant ALK) and TRKA TRKB and TRKC. purity: 99%