Learn More
Medchemexpress LLC HY-17603 5mg Medchemexpress, Belizatinib CAS:1357920-84-3 Purity:>98%
SDP
Catalog No. 502082000
Encompass_Preferred
Medchemexpress, HY-17603 5mg Belizatinib CAS:1357920-84-3 Belizatinib is an oral, dual, potent inhibitor of ALK and TRKA, TRKB, and TRKC, with IC50 of 0.7 nM for wild-type recombinant ALK kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2000 Supplier Medchemexpress LLC Supplier No. HY176035MG
Medchemexpress, HY-17603 5mg Belizatinib CAS:1357920-84-3 Belizatinib is an oral, dual, potent inhibitor of ALK and TRKA, TRKB, and TRKC, with IC50 of 0.7 nM for wild-type recombinant ALK kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.