Learn More
Enterprise Technology Solutions Belkin CAT6 Snagless Patch Cable, 15' L, Blue
SDP
Catalog No. 5000739545
Encompass_Preferred
RJ45 High-Performance Category 6 Snagless Patch Cable offers high bandwidth to accommodate streaming video and other data-intensive applications. Ideal for 100 Base-T and gigabit networks. Performance-tested to 100 MHz frequency. Cable provides the highest-performance network connection. Patch cable increases the quality of high-speed Internet applications, multiplayer gaming and streaming audio video. Ideal for demanding use and small/medium-size business.
Catalog No. 50-007-39545 Supplier Enterprise Technology Solutions Supplier No. BLKA3L98015BLUS
RJ45 High-Performance Category 6 Snagless Patch Cable offers high bandwidth to accommodate streaming video and other data-intensive applications. Ideal for 100 Base-T and gigabit networks. Performance-tested to 100 MHz frequency. Cable provides the highest-performance network connection. Patch cable increases the quality of high-speed Internet applications, multiplayer gaming and streaming audio video. Ideal for demanding use and small/medium-size business.