Learn More
Enterprise Technology Solutions Belkin Networking Cat6 Bulk Cable, Stranded, 1000', Black
SDP
Supplier:  Enterprise Technology Solutions BLKA7J7041000BK
Encompass_Preferred
CAT6 stranded cable features a 1,000' length to let you build your cables to your exact specifications and eliminate excess cable. CAT6 technology offers a very high bandwidth to maximize your network speed when working on data-intensive applications. 24AWG wires are performance tested to exceed 500MHz for clean, clear transmissions. CAT6 cable is perfect for high-end solutions such as Gigabit Ethernet, 1.2Gbps ATM and broadband video.
Catalog No. 50-007-39590